siRNA Details
| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199391.1 | NP_955423.1 | 11, 12 | 1657 | ||
| XM_008768572.1 | XP_008766794.1 | 11, 12 | 1764 | |||
| XM_008768573.1 | XP_008766795.1 | 11, 12 | 1761 | |||
| XM_008768574.1 | XP_008766796.1 | 11, 12 | 1767 | |||
| XM_008768575.1 | XP_008766797.1 | 11, 12 | 1767 | |||
| XM_008768576.1 | XP_008766798.1 | 12, 13 | 1366 | |||
| XM_008768577.1 | XP_008766799.1 | 11, 12 | 1241 | |||
| XM_008768578.1 | XP_008766800.1 | 11, 12 | 1604 | |||
| GenBank mRNA | BC062389.1 | - | 11, 12 | 1657 | 5620907 | |

C2cd2
Genomic Map