siRNA Details
| Gene Symbol: Cbr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107110.1 | NP_001100580.1 | 3 | 772 | ||
| GenBank mRNA | BC166553.1 | - | 3 | 733 | 7310545 | |
| FQ234106.1 | - | 3 | 750 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbr3| Gene Symbol: Cbr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107110.1 | NP_001100580.1 | 3 | 772 | ||
| GenBank mRNA | BC166553.1 | - | 3 | 733 | 7310545 | |
| FQ234106.1 | - | 3 | 750 | |||