siRNA Details
| Gene Symbol: Cldn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037774.1 | NP_001032863.1 | 1 | 589 | ||
| GenBank mRNA | BC099804.1 | - | 1 | 589 | 7937572 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn8| Gene Symbol: Cldn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037774.1 | NP_001032863.1 | 1 | 589 | ||
| GenBank mRNA | BC099804.1 | - | 1 | 589 | 7937572 | |