siRNA Details
| Gene Symbol: Rwdd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100559.1 | NP_001094029.1 | 5 | 797 | ||
| GenBank mRNA | BC091390.1 | - | 1 | 68 | ||
| BC161911.1 | - | 5 | 797 | 8368806 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rwdd2b| Gene Symbol: Rwdd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100559.1 | NP_001094029.1 | 5 | 797 | ||
| GenBank mRNA | BC091390.1 | - | 1 | 68 | ||
| BC161911.1 | - | 5 | 797 | 8368806 | ||