siRNA Details
Gene Symbol: Adamts5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198761.1 | NP_942056.1 | 6 | 2025 | ||
GenBank mRNA | AY382879.1 | - | 6 | 2025 | ||
FQ228283.1 | - | 2418 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adamts5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198761.1 | NP_942056.1 | 6 | 2025 | ||
GenBank mRNA | AY382879.1 | - | 6 | 2025 | ||
FQ228283.1 | - | 2418 |