siRNA Details
| Gene Symbol: MGC95208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005552.1 | NP_001005552.1 | 3 | 472 | ||
| GenBank mRNA | BC082055.1 | - | 3 | 472 | 7132244 | |
| BC091244.1 | - | 3 | 467 | 7365330 | ||
| Gene Symbol: LOC100362724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752027.2 | XP_003752075.1 | 1 | 462 | ||

MGC95208
Genomic Map