siRNA Details
| Gene Symbol: Lrrc8e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034139.1 | NP_001029311.1 | 3 | 443 | ||
| XM_006248763.2 | XP_006248825.1 | 3 | 479 | |||
| XM_006248764.2 | XP_006248826.1 | 4 | 612 | |||
| XM_008768954.1 | XP_008767176.1 | 3 | 747 | |||
| XM_008768955.1 | XP_008767177.1 | 2 | 276 | |||
| GenBank mRNA | BC105779.1 | - | 3 | 443 | 7932319 | |

Lrrc8e
Genomic Map