siRNA Details
| Gene Symbol: Pds5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101805.1 | NP_001095275.1 | 25 | 2980 | ||
| NM_001102383.2 | NP_001095853.1 | 25 | 2980 | |||
| XM_008768957.1 | XP_008767179.1 | 26 | 3066 | |||
| XM_008768959.1 | XP_008767181.1 | 26 | 3009 | |||

Pds5b
Genomic Map