siRNA Details
| Gene Symbol: Pds5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101805.1 | NP_001095275.1 | 20 | 2402 | ||
| NM_001102383.2 | NP_001095853.1 | 20 | 2402 | |||
| XM_008768957.1 | XP_008767179.1 | 21 | 2488 | |||
| XM_008768958.1 | XP_008767180.1 | 21 | 2519 | |||
| XM_008768959.1 | XP_008767181.1 | 21 | 2431 | |||

Pds5b
Genomic Map