siRNA Details
| Gene Symbol: Vom2r59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099466.1 | NP_001092936.1 | 3 | 672 | ||
| Gene Symbol: Vom2r58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099479.1 | NP_001092949.1 | 3 | 672 | ||
More Information
Set Membership: |
|

Vom2r59
Genomic Map