siRNA Details
| Gene Symbol: Tmem130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170399.1 | NP_001163870.1 | 4 | 792 | ||
| GenBank mRNA | BP484923.1 | - | 1 | 59 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem130| Gene Symbol: Tmem130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170399.1 | NP_001163870.1 | 4 | 792 | ||
| GenBank mRNA | BP484923.1 | - | 1 | 59 | ||