siRNA Details
| Gene Symbol: Baiap2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034140.1 | NP_001029312.1 | 5 | 559 | ||
| GenBank mRNA | BC089216.1 | - | 4 | 402 | ||
| BC105815.1 | - | 5 | 559 | 7367101 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baiap2l1| Gene Symbol: Baiap2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034140.1 | NP_001029312.1 | 5 | 559 | ||
| GenBank mRNA | BC089216.1 | - | 4 | 402 | ||
| BC105815.1 | - | 5 | 559 | 7367101 | ||