siRNA Details
| Gene Symbol: Bri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009604.1 | NP_001009604.1 | 3 | 533 | ||
| GenBank mRNA | BC087646.1 | - | 3 | 533 | 7311234 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bri3| Gene Symbol: Bri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009604.1 | NP_001009604.1 | 3 | 533 | ||
| GenBank mRNA | BC087646.1 | - | 3 | 533 | 7311234 | |