siRNA Details
| Gene Symbol: Kdelr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013122.1 | NP_001013140.1 | 2, 3 | 291 | ||
| GenBank mRNA | BC085786.1 | - | 2, 3 | 291 | 7108608 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelr2| Gene Symbol: Kdelr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013122.1 | NP_001013140.1 | 2, 3 | 291 | ||
| GenBank mRNA | BC085786.1 | - | 2, 3 | 291 | 7108608 | |