siRNA Details
| Gene Symbol: Sdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769059.1 | XP_008767281.1 | 19, 20 | 2751 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdk1| Gene Symbol: Sdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769059.1 | XP_008767281.1 | 19, 20 | 2751 | ||