siRNA Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107123.1 | NP_001100593.1 | 15 | 5598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnrc18| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107123.1 | NP_001100593.1 | 15 | 5598 | ||