siRNA Details
| Gene Symbol: Ttyh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107124.1 | NP_001100594.1 | 9 | 973 | ||
| XM_008768969.1 | XP_008767191.1 | 9 | 1162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttyh3| Gene Symbol: Ttyh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107124.1 | NP_001100594.1 | 9 | 973 | ||
| XM_008768969.1 | XP_008767191.1 | 9 | 1162 | |||