siRNA Details
| Gene Symbol: Zfp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107128.2 | NP_001100598.2 | 6 | 1921 | ||
| NM_001142945.1 | NP_001136417.1 | 5 | 1825 | |||
| XM_008768974.1 | XP_008767196.1 | 3 | 2290 | |||
| XM_008768975.1 | XP_008767197.1 | 6 | 1979 | |||

Zfp68
Genomic Map