siRNA Details
| Gene Symbol: Ap4m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037977.1 | NP_001033066.1 | 10 | 759 | ||
| XM_006249008.2 | XP_006249070.1 | 10 | 788 | |||
| GenBank mRNA | BC127443.1 | - | 10 | 824 | ||
| BC161814.1 | - | 11 | 953 | |||
| DQ298439.1 | - | 10 | 759 | |||

Ap4m1
Genomic Map