siRNA Details
| Gene Symbol: Fbxo24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107130.2 | NP_001100600.2 | 6 | 928 | ||
| XM_006249139.2 | XP_006249201.1 | 6 | 1441 | |||
| XM_008769135.1 | XP_008767357.1 | 6 | 1510 | |||
| XM_008769136.1 | XP_008767358.1 | 7 | 1532 | |||

Fbxo24
Genomic Map