siRNA Details
| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107132.1 | NP_001100602.1 | 4 | 1405 | ||
| XM_006249143.2 | XP_006249205.1 | 1955 | ||||
| XM_006249144.2 | XP_006249206.1 | 4 | 1597 | |||
| XM_006249145.2 | XP_006249207.1 | 4 | 1265 | |||
| XM_006249147.2 | XP_006249209.1 | 4 | 1005 | |||
| XM_006249149.2 | XP_006249211.1 | 1955 | ||||
| Gene Symbol: LOC100910838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751152.3 | XP_003751200.2 | 4 | 1597 | ||
| XM_006249084.2 | XP_006249146.1 | 3 | 1956 | |||
| XM_006249085.2 | XP_006249147.1 | 4 | 1266 | |||
| XM_006249087.2 | XP_006249149.1 | 4 | 1006 | |||
| XM_006249089.2 | XP_006249151.1 | 3 | 1955 | |||
| Gene Symbol: LOC103691013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758869.1 | XP_008757091.1 | 2 | 1255 | ||

Nyap1
Genomic Map