siRNA Details
| Gene Symbol: LOC304396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013981.1 | NP_001014003.1 | 2 | 349 | ||
| XM_006249156.2 | XP_006249218.1 | 2 | 402 | |||
| GenBank mRNA | BC081940.1 | - | 2 | 349 | 7115744 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC304396| Gene Symbol: LOC304396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013981.1 | NP_001014003.1 | 2 | 349 | ||
| XM_006249156.2 | XP_006249218.1 | 2 | 402 | |||
| GenBank mRNA | BC081940.1 | - | 2 | 349 | 7115744 | |