siRNA Details
| Gene Symbol: Ssc4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249209.2 | XP_006249271.1 | 8 | 1407 | ||
| XM_006249211.2 | XP_006249273.1 | 8 | 1407 | |||
| XM_222076.8 | XP_222076.5 | 8 | 1372 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssc4d| Gene Symbol: Ssc4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249209.2 | XP_006249271.1 | 8 | 1407 | ||
| XM_006249211.2 | XP_006249273.1 | 8 | 1407 | |||
| XM_222076.8 | XP_222076.5 | 8 | 1372 | |||