siRNA Details
| Gene Symbol: SSPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135823.1 | NP_001129295.1 | 3 | 446 | ||
| NM_005086.4 | NP_005077.2 | 3 | 629 | |||
| XM_011520853.2 | XP_011519155.1 | 3 | 379 | |||
| XM_011520854.2 | XP_011519156.1 | 3 | 553 | |||
| XM_011520855.2 | XP_011519157.1 | 3 | 295 | |||
| GenBank mRNA | AF016028.2 | - | 3 | 529 | ||
| AK092925.1 | - | 3 | 442 | |||
| AK304667.1 | - | 4 | 523 | |||
| AK314353.1 | - | 3 | 618 | |||
| AL136756.1 | - | 3 | 613 | |||
| BC062299.1 | - | 3 | 609 | 6376868 | ||
| DA073350.1 | - | 4 | 414 | |||
| X89105.1 | - | 3 | 451 | |||

SSPN
Genomic Map