siRNA Details
| Gene Symbol: Glt1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769205.1 | XP_008767427.1 | 3 | 529 | ||
| XM_222147.8 | XP_222147.4 | 3 | 384 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glt1d1| Gene Symbol: Glt1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769205.1 | XP_008767427.1 | 3 | 529 | ||
| XM_222147.8 | XP_222147.4 | 3 | 384 | |||