siRNA Details
| Gene Symbol: Rilpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191665.1 | NP_001178594.1 | 5, 6 | 1188 | ||
| XM_006249294.1 | XP_006249356.1 | 5, 6 | 1452 | |||
| XM_006249295.1 | XP_006249357.1 | 5, 6 | 1452 | |||
| XM_006249296.2 | XP_006249358.1 | 5, 6 | 1452 | |||
| XM_006249297.2 | XP_006249359.1 | 5, 6 | 1452 | |||

Rilpl1
Genomic Map