siRNA Details
| Gene Symbol: Kntc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107140.1 | NP_001100610.1 | 44 | 5012 | ||
| XM_008769228.1 | XP_008767450.1 | 50 | 5598 | |||
| XM_008769229.1 | XP_008767451.1 | 50 | 5418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kntc1| Gene Symbol: Kntc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107140.1 | NP_001100610.1 | 44 | 5012 | ||
| XM_008769228.1 | XP_008767450.1 | 50 | 5598 | |||
| XM_008769229.1 | XP_008767451.1 | 50 | 5418 | |||