siRNA Details
| Gene Symbol: Ccdc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025665.1 | NP_001020836.1 | 10 | 1615 | ||
| GenBank mRNA | BC097409.1 | - | 10 | 1615 | 7454160 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc63| Gene Symbol: Ccdc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025665.1 | NP_001020836.1 | 10 | 1615 | ||
| GenBank mRNA | BC097409.1 | - | 10 | 1615 | 7454160 | |