siRNA Details
| Gene Symbol: Kdm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100679.1 | NP_001094149.1 | 19 | 3018 | ||
| GenBank mRNA | BC082040.1 | - | 1 | 48 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdm2b| Gene Symbol: Kdm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100679.1 | NP_001094149.1 | 19 | 3018 | ||
| GenBank mRNA | BC082040.1 | - | 1 | 48 | ||