siRNA Details
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107144.1 | NP_001100614.1 | 4 | 610 | ||
XM_008769232.1 | XP_008767454.1 | 4 | 571 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107144.1 | NP_001100614.1 | 4 | 610 | ||
XM_008769232.1 | XP_008767454.1 | 4 | 571 |