siRNA Details
| Gene Symbol: Rfc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107146.2 | NP_001100616.1 | 7 | 668 | ||
| GenBank mRNA | BC168902.1 | - | 7 | 668 | 7388155 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rfc5| Gene Symbol: Rfc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107146.2 | NP_001100616.1 | 7 | 668 | ||
| GenBank mRNA | BC168902.1 | - | 7 | 668 | 7388155 | |