siRNA Details
| Gene Symbol: Coq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039022.1 | NP_001034111.1 | 3 | 469 | ||
| GenBank mRNA | BC098719.1 | - | 3 | 469 | 7370143 | |
| FQ219423.1 | - | 3 | 474 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq5| Gene Symbol: Coq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039022.1 | NP_001034111.1 | 3 | 469 | ||
| GenBank mRNA | BC098719.1 | - | 3 | 469 | 7370143 | |
| FQ219423.1 | - | 3 | 474 | |||