siRNA Details
| Gene Symbol: Git2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005553.1 | NP_001005553.1 | 20 | 2306 | ||
| XM_006249442.2 | XP_006249504.1 | 20 | 2351 | |||
| XM_006249445.2 | XP_006249507.1 | 19 | 2264 | |||
| XM_006249447.2 | XP_006249509.1 | 19 | 2201 | |||
| XM_006249450.2 | XP_006249512.1 | 18 | 2114 | |||
| XM_008769324.1 | XP_008767546.1 | 20 | 2309 | |||
| XM_008769325.1 | XP_008767547.1 | 19 | 2204 | |||
| GenBank mRNA | BC081967.1 | - | 20 | 2306 | 7125406 | |

Git2
Genomic Map