siRNA Details
| Gene Symbol: Myo18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249577.2 | XP_006249639.1 | 6, 7 | 2046 | ||
| XM_006249578.2 | XP_006249640.1 | 5, 6 | 1835 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo18b| Gene Symbol: Myo18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249577.2 | XP_006249639.1 | 6, 7 | 2046 | ||
| XM_006249578.2 | XP_006249640.1 | 5, 6 | 1835 | |||