siRNA Details
| Gene Symbol: Pus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025563.1 | NP_001020734.1 | 5 | 1376 | ||
| XM_006249547.2 | XP_006249609.1 | 5 | 1513 | |||
| XM_008769330.1 | XP_008767552.1 | 5 | 1521 | |||
| XM_008769331.1 | XP_008767553.1 | 5 | 1517 | |||
| GenBank mRNA | BC161821.1 | - | 5 | 1336 | 9027947 | |

Pus1
Genomic Map