siRNA Details
| Gene Symbol: Ddx51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107150.1 | NP_001100620.1 | 10, 11 | 1497 | ||
| XM_006249561.2 | XP_006249623.1 | 10, 11 | 1638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx51| Gene Symbol: Ddx51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107150.1 | NP_001100620.1 | 10, 11 | 1497 | ||
| XM_006249561.2 | XP_006249623.1 | 10, 11 | 1638 | |||