siRNA Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249589.2 | XP_006249651.1 | 9 | 1646 | ||
| XM_006249592.2 | XP_006249654.1 | 9 | 1647 | |||
| XM_008769388.1 | XP_008767610.1 | 9 | 1644 | |||
| XM_008769389.1 | XP_008767611.1 | 8 | 1616 | |||
| XM_222254.8 | XP_222254.5 | 10 | 1716 | |||

Fbrsl1
Genomic Map