siRNA Details
| Gene Symbol: Pole | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107152.2 | NP_001100622.2 | 21 | 2499 | ||
| XM_006249562.2 | XP_006249624.1 | 21 | 2536 | |||
| XM_008769335.1 | XP_008767557.1 | 21 | 2536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pole| Gene Symbol: Pole | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107152.2 | NP_001100622.2 | 21 | 2499 | ||
| XM_006249562.2 | XP_006249624.1 | 21 | 2536 | |||
| XM_008769335.1 | XP_008767557.1 | 21 | 2536 | |||