siRNA Details
| Gene Symbol: Plcxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249597.2 | XP_006249659.1 | 124 | |||
| Gene Symbol: LOC100909600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249594.2 | XP_006249656.1 | 2 | 124 | ||

Plcxd1
Genomic Map