siRNA Details
| Gene Symbol: Ung | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013124.1 | NP_001013142.1 | 4 | 602 | ||
| XM_008769336.1 | XP_008767558.1 | 5 | 1273 | |||
| GenBank mRNA | BC091211.1 | - | 4 | 602 | 7381643 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ung| Gene Symbol: Ung | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013124.1 | NP_001013142.1 | 4 | 602 | ||
| XM_008769336.1 | XP_008767558.1 | 5 | 1273 | |||
| GenBank mRNA | BC091211.1 | - | 4 | 602 | 7381643 | |