siRNA Details
Gene Symbol: Baz2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107158.1 | NP_001100628.1 | 26 | 5232 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Baz2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107158.1 | NP_001100628.1 | 26 | 5232 |