siRNA Details
| Gene Symbol: Olr1065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000498.1 | NP_001000498.1 | 1 | 446 | ||
| Gene Symbol: LOC100910887 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751221.2 | XP_003751269.2 | 1 | 446 | ||
| Gene Symbol: Olr1063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000064.1 | NP_001000064.1 | 1 | 446 | ||

Olr1065
Genomic Map