siRNA Details
| Gene Symbol: Tbc1d9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134539.1 | NP_001128011.1 | 6 | 1598 | ||
| XM_006255246.2 | XP_006255308.1 | 6 | 2239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d9| Gene Symbol: Tbc1d9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134539.1 | NP_001128011.1 | 6 | 1598 | ||
| XM_006255246.2 | XP_006255308.1 | 6 | 2239 | |||