siRNA Details
Gene Symbol: Mgat4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025666.1 | NP_001020837.1 | 7, 8 | 832 | ||
XM_008772379.1 | XP_008770601.1 | 7, 8 | 917 | |||
XM_008772380.1 | XP_008770602.1 | 7, 8 | 769 | |||
GenBank mRNA | BC097408.1 | - | 7, 8 | 832 | 7454086 |
