siRNA Details
Gene Symbol: Mgat4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025666.1 | NP_001020837.1 | 1, 2 | 169 | ||
GenBank mRNA | BC097408.1 | - | 1, 2 | 169 | 7454086 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgat4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025666.1 | NP_001020837.1 | 1, 2 | 169 | ||
GenBank mRNA | BC097408.1 | - | 1, 2 | 169 | 7454086 |