siRNA Details
| Gene Symbol: Asf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107160.1 | NP_001100630.1 | 3 | 494 | ||
| GenBank mRNA | BC158705.1 | - | 3 | 420 | 9030040 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asf1b| Gene Symbol: Asf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107160.1 | NP_001100630.1 | 3 | 494 | ||
| GenBank mRNA | BC158705.1 | - | 3 | 420 | 9030040 | |