siRNA Details
| Gene Symbol: Zswim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107163.1 | NP_001100633.1 | 14 | 2502 | ||
| XM_006255251.2 | XP_006255313.1 | 14 | 2541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim4| Gene Symbol: Zswim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107163.1 | NP_001100633.1 | 14 | 2502 | ||
| XM_006255251.2 | XP_006255313.1 | 14 | 2541 | |||