siRNA Details
| Gene Symbol: Lyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007677.1 | NP_001007678.1 | 4 | 1414 | ||
| GenBank mRNA | BC081864.1 | - | 4 | 1414 | 7096152 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyl1| Gene Symbol: Lyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007677.1 | NP_001007678.1 | 4 | 1414 | ||
| GenBank mRNA | BC081864.1 | - | 4 | 1414 | 7096152 | |