siRNA Details
Gene Symbol: Epb4.1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012023.1 | NP_001012023.1 | 12 | 1149 | ||
GenBank mRNA | BC090012.1 | - | 12 | 1149 | 7324591 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Epb4.1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012023.1 | NP_001012023.1 | 12 | 1149 | ||
GenBank mRNA | BC090012.1 | - | 12 | 1149 | 7324591 |