siRNA Details
| Gene Symbol: Ccdc93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024997.1 | NP_001020168.1 | 18, 19 | 1542 | ||
| XM_008769456.1 | XP_008767678.1 | 19, 20 | 1576 | |||
| GenBank mRNA | BC091337.1 | - | 18, 19 | 1542 | 7314934 | |

Ccdc93
Genomic Map